ENA336956076 Enamine

2-hydroxy-5-{[(propan-2-yl)carbamoyl]amino}benzoic acid

  • MDL number MFCD11167440
  • Cas Number : 1038255-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84966
Availability details for
Quantity
  • Availability
  • Add to Cart