ENA336956078 Enamine

[4-(2,3-dichlorophenoxy)phenyl]methanamine

  • MDL number MFCD11191753
  • Cas Number : 1039894-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84971

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84971
Availability details for
Quantity
  • Availability
  • Add to Cart