ENA336956079 Enamine

N,1,2-trimethylpiperidin-4-amine

  • MDL number MFCD14692594
  • Cas Number : 90203-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84973

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84973
Availability details for
Quantity
  • Availability
  • Add to Cart