ENA336956081 Enamine

2-(2,3-dimethylphenyl)pyrrolidine

  • MDL number MFCD05189441
  • Cas Number : 899365-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84975
Availability details for
Quantity
  • Availability
  • Add to Cart