ENA336956096 Enamine

1-[4-(2-methoxyphenyl)phenyl]ethan-1-one

  • MDL number MFCD04039104
  • Cas Number : 52807-17-7
  • Molecular Weight : 226.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-85005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85005
Availability details for
Quantity
  • Availability
  • Add to Cart