ENA336956104 Enamine

2-(aminomethyl)-N,N-dimethyl-2,3-dihydro-1H-inden-2-amine

  • MDL number MFCD12067386
  • Cas Number : 1157497-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85024
Availability details for
Quantity
  • Availability
  • Add to Cart