ENA336956109 Enamine

4-chloro-6-fluoro-8-methylquinoline-3-carbonitrile

  • MDL number MFCD09934836
  • Cas Number : 1016773-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85033
Availability details for
Quantity
  • Availability
  • Add to Cart