ENA336956118 Enamine

2-(2-hydroxyethoxy)pyridine-3-carboxylic acid

  • MDL number MFCD11136680
  • Cas Number : 672957-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196...198
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85043

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85043
Availability details for
Quantity
  • Availability
  • Add to Cart