ENA336956124 Enamine

2-bromo-3-methyl-1-(1,2,3,4-tetrahydroisoquinolin-2-yl)butan-1-one

  • MDL number MFCD09949239
  • Cas Number : 1017030-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29137
Availability details for
Quantity
  • Availability
  • Add to Cart