ENA336956129 Enamine

2-bromo-3-methyl-1-(2-methylpiperidin-1-yl)butan-1-one

  • MDL number MFCD09932520
  • Cas Number : 81042-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29155
Availability details for
Quantity
  • Availability
  • Add to Cart