ENA336956133 Enamine

4-methoxy-3-(1H-1,2,3,4-tetrazol-1-yl)aniline

  • MDL number MFCD09941394
  • Cas Number : 891017-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85090

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85090
Availability details for
Quantity
  • Availability
  • Add to Cart