ENA336956137 Enamine

benzyl(2-methylpropyl)amine

  • MDL number MFCD00972061
  • Cas Number : 42882-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32577
Availability details for
Quantity
  • Availability
  • Add to Cart