ENA336956141 Enamine

3-amino-1-(2-chlorophenyl)piperidin-2-one

  • MDL number MFCD19679255
  • Cas Number : 1341769-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85100
Availability details for
Quantity
  • Availability
  • Add to Cart