ENA336956142 Enamine

3-(pentan-3-yl)-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD16706799
  • Cas Number : 1248155-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85101
Availability details for
Quantity
  • Availability
  • Add to Cart