ENA336956144 Enamine

1-phenylhexan-3-amine

  • MDL number MFCD11655387
  • Cas Number : 5391-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85103
Availability details for
Quantity
  • Availability
  • Add to Cart