ENA336956147 Enamine

3-(pentan-3-yl)-1H-pyrazol-5-amine

  • MDL number MFCD10692133
  • Cas Number : 81114-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 80
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85107

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85107
Availability details for
Quantity
  • Availability
  • Add to Cart