ENA336956155 Enamine

3-[2-(trifluoromethyl)phenyl]butanoic acid

  • MDL number MFCD11172900
  • Cas Number : 1042815-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81899
Availability details for
Quantity
  • Availability
  • Add to Cart