ENA336956161 Enamine

5-[({2-[4-(propan-2-yl)phenyl]-1,3-thiazol-5-yl}methyl)sulfanyl]-4H-1,2,4-triazol-3-amine

  • MDL number MFCD06376325
  • Cas Number : 851213-82-6
  • Molecular Weight : 331.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82188
Availability details for
Quantity
  • Availability
  • Add to Cart