ENA336956164 Enamine

3-hydroxy-6-methoxy-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD14641541
  • Cas Number : 1248148-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85149

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85149
Availability details for
Quantity
  • Availability
  • Add to Cart