ENA336956165 Enamine

2-azabicyclo[2.2.1]heptane-2-sulfonyl chloride

  • MDL number MFCD14693843
  • Cas Number : 1248803-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85150
Availability details for
Quantity
  • Availability
  • Add to Cart