ENA336956170 Enamine

2-[(2,2-difluoroethyl)sulfanyl]-5-(trifluoromethyl)aniline

  • MDL number MFCD12807580
  • Cas Number : 1183293-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82289

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82289
Availability details for
Quantity
  • Availability
  • Add to Cart