ENA336956171 Enamine

3-(4-aminophenyl)-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD14631612
  • Cas Number : 62036-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 333...335
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85162

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85162
Availability details for
Quantity
  • Availability
  • Add to Cart