ENA336956172 Enamine

1-(3-bromophenyl)-1H,4H,5H,6H-cyclopenta[c]pyrazole-3-carboxylic acid

  • MDL number MFCD09048680
  • Cas Number : 926234-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85164

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-85164
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart