ENA336956173 Enamine

3-(aminomethyl)-1-benzylpyrrolidin-3-ol

  • MDL number MFCD12169429
  • Cas Number : 125033-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82290
Availability details for
Quantity
  • Availability
  • Add to Cart