ENA336956174 Enamine

2-(cyclohexyloxy)benzoic acid

  • MDL number MFCD11156584
  • Cas Number : 55751-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86...88
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85166

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85166
Availability details for
Quantity
  • Availability
  • Add to Cart