ENA336956176 Enamine

5-methoxypentan-1-ol

  • MDL number MFCD00046025
  • Cas Number : 4799-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85168

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85168
Availability details for
Quantity
  • Availability
  • Add to Cart