ENA336956178 Enamine

propan-2-yl 3-chloropropanoate

  • MDL number MFCD00018977
  • Cas Number : 691-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-85170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85170
Availability details for
Quantity
  • Availability
  • Add to Cart