ENA336956179 Enamine

[2-({[(furan-2-yl)methyl](methyl)amino}methyl)phenyl]methanamine

  • MDL number MFCD12426533
  • Cas Number : 1155480-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85171
Availability details for
Quantity
  • Availability
  • Add to Cart