ENA336956185 Enamine

[2-(4-methylpiperazin-1-yl)-5-(trifluoromethyl)phenyl]methanamine

  • MDL number MFCD12652176
  • Cas Number : 1094329-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85184

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-85184
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart