ENA336956189 Enamine

2-(N-ethyl2-chloropyridine-3-sulfonamido)acetic acid

  • MDL number MFCD12904751
  • Cas Number : 1183511-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85227
Availability details for
Quantity
  • Availability
  • Add to Cart