ENA336956193 Enamine

6-amino-N-ethyl-2,3-dihydro-1H-indole-1-carboxamide

  • MDL number MFCD11106242
  • Cas Number : 1152893-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85233
Availability details for
Quantity
  • Availability
  • Add to Cart