ENA336956200 Enamine

(2-chlorophenyl)(4-ethoxyphenyl)methanone

  • MDL number MFCD00687607
  • Cas Number : 525561-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86...88
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85260

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85260
Availability details for
Quantity
  • Availability
  • Add to Cart