ENA336956203 Enamine

methyl 5-amino-4-bromo-2-fluorobenzoate

  • MDL number MFCD17168715
  • Cas Number : 1342446-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85266
Availability details for
Quantity
  • Availability
  • Add to Cart