ENA336956205 Enamine

3-(2-methyl-1,2,3,4-tetrahydroquinolin-1-yl)propanoic acid

  • MDL number MFCD11117476
  • Cas Number : 1036563-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85272

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85272
Availability details for
Quantity
  • Availability
  • Add to Cart