ENA336956209 Enamine

[2-(piperazin-1-yl)ethyl](propan-2-yl)amine

  • MDL number MFCD12796605
  • Cas Number : 1307634-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85276
Availability details for
Quantity
  • Availability
  • Add to Cart