ENA336956211 Enamine

2-amino-2-(3-methoxyphenyl)ethan-1-ol

  • MDL number MFCD11213674
  • Cas Number : 325153-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85278
Availability details for
Quantity
  • Availability
  • Add to Cart