ENA336956212 Enamine

1-(2-chlorophenyl)piperidin-3-amine

  • MDL number MFCD16074671
  • Cas Number : 1250430-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85280

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85280
Availability details for
Quantity
  • Availability
  • Add to Cart