ENA336956213 Enamine

[1,3-bis(4-methylphenyl)propan-2-yl](methyl)amine

  • MDL number MFCD17960702
  • Cas Number : 1282602-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85281

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85281
Availability details for
Quantity
  • Availability
  • Add to Cart