ENA336956214 Enamine

1,3-bis(4-methylphenyl)propan-2-amine

  • MDL number MFCD09887265
  • Cas Number : 74548-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85282
Availability details for
Quantity
  • Availability
  • Add to Cart