ENA336956218 Enamine

5-tert-butyl-1-[(2-fluorophenyl)methyl]-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD19269628
  • Cas Number : 1266832-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85290
Availability details for
Quantity
  • Availability
  • Add to Cart