ENA336956223 Enamine

2-(4-bromo-2-fluorophenyl)-2-chloroacetamide

  • MDL number MFCD16694394
  • Cas Number : 1249610-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82580
Availability details for
Quantity
  • Availability
  • Add to Cart