ENA336956234 Enamine

3-amino-4-methoxy-N-(pyridin-3-yl)benzene-1-sulfonamide

  • MDL number MFCD12737276
  • Cas Number : 1041546-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85349
Availability details for
Quantity
  • Availability
  • Add to Cart