ENA336956241 Enamine

2-(2-isocyanatopropyl)thiophene

  • MDL number MFCD11612970
  • Cas Number : 1082916-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85367
Availability details for
Quantity
  • Availability
  • Add to Cart