ENA336956249 Enamine

1,2-dichloro-3-[4-(chloromethyl)phenoxy]benzene

  • MDL number MFCD11190593
  • Cas Number : 1038975-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85410
Availability details for
Quantity
  • Availability
  • Add to Cart