ENA336956252 Enamine

(1S,3S,5S)-2-azabicyclo[3.1.0]hexane-3-carboxylic acid hydrochloride

  • MDL number MFCD09971685
  • Cas Number : 1807937-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39001

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39001
Availability details for
Quantity
  • Availability
  • Add to Cart