ENA336956257 Enamine

2-(2,6-difluorophenyl)-2-methylcyclopropane-1-carboxylic acid

  • MDL number MFCD16744571
  • Cas Number : 1250566-64-3
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85419
Availability details for
Quantity
  • Availability
  • Add to Cart