ENA336956258 Enamine

1-[5-methyl-1-(quinolin-5-yl)-1H-1,2,3-triazol-4-yl]ethan-1-one

  • MDL number MFCD12592226
  • Cas Number : 1157067-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85420
Availability details for
Quantity
  • Availability
  • Add to Cart