ENA336956264 Enamine

1-methyl-1H,2H,3H-imidazo[4,5-c]pyridin-2-one

  • MDL number MFCD18258912
  • Cas Number : 40423-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85428
Availability details for
Quantity
  • Availability
  • Add to Cart