ENA336956266 Enamine

4-(4-isocyanatophenyl)morpholine

  • MDL number MFCD09608103
  • Cas Number : 884332-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85430

Safety Information

signalword Danger
pcodes P261,P264,P270,P280,P285,P301+P310,P304+P341,P305+P351+P338,P310,P330,P342+P311,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-85430
pictograms GHS05,GHS06,GHS08
hstatements H301,H318,H334
Availability details for
Quantity
  • Availability
  • Add to Cart