ENA336956271 Enamine

tert-butyl 2-(phenylamino)acetate

  • MDL number MFCD11848799
  • Cas Number : 65171-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85435
Availability details for
Quantity
  • Availability
  • Add to Cart